CH6371377917 Chem Impex International, Inc.

Nim-Trityl-histidine

  • MDL number MFCD00153444
  • Cas Number : 35146-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point ~ 200
Synonym L-His(Trt)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-trityl-histidine/1989?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-trityl-histidine/1989?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart