CH6371377918 Chem Impex International, Inc.

Fmoc-b-cyclohexyl-D-alanine

  • MDL number MFCD00151912
  • Cas Number : 144701-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Cha-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-ß-cyclohexyl-d-alanine/1990?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-ß-cyclohexyl-d-alanine/1990?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart