CH6371377929 Chem Impex International, Inc.

Benzoyl-L-alanine methyl ester

  • MDL number MFCD00025871
  • Cas Number : 7244-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Bz-L-Ala-OMe
Storage temp. Store at 0-8
Physical Form Crystalline
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzoyl-l-alanine-methyl-ester/2002?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzoyl-l-alanine-methyl-ester/2002?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart