CH6371377933 Chem Impex International, Inc.

D-Arginine methyl ester dihydrochloride

  • MDL number MFCD00153417
  • Cas Number : 78851-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-184� C
Synonym D-Arg-OMe 2HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-arginine-methyl-ester-dihydrochloride/2006?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-arginine-methyl-ester-dihydrochloride/2006?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart