CH6371377937 Chem Impex International, Inc.

Nw-Nitro-D-arginine methyl ester hydrochloride

  • MDL number MFCD00133613
  • Cas Number : 50912-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-164 �C
Synonym D-Arg(NO2)-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White to off white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-omega-sup-nitro-d-arginine-methyl-ester-hydrochloride/2010?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-omega-sup-nitro-d-arginine-methyl-ester-hydrochloride/2010?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart