CH6371377941 Chem Impex International, Inc.

Nalpha-Benzoyl-L-arginine methyl ester hydrochloride

  • MDL number MFCD00038945
  • Cas Number : 1784-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-benzoyl-l-arginine-methyl-ester-hydrochloride/2015?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-benzoyl-l-arginine-methyl-ester-hydrochloride/2015?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart