CH6371377948 Chem Impex International, Inc.

L-Aspartic acid beta-benzyl ester alpha-amide hydrochloride

  • MDL number MFCD00153425
  • Cas Number : 199118-68-8
  • Molecular Weight : 258.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186-196 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-aspartic-acid-ß-benzyl-ester-a-amide-hydrochloride/2022?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-aspartic-acid-ß-benzyl-ester-a-amide-hydrochloride/2022?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart