CH6371377951 Chem Impex International, Inc.

S-Acetamidomethyl-L-cysteine hydrochloride

  • MDL number MFCD00077080
  • Cas Number : 28798-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-acetamidomethyl-l-cysteine-hydrochloride/2026?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-acetamidomethyl-l-cysteine-hydrochloride/2026?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart