CH6371377953 Chem Impex International, Inc.

S-Benzyl-D-cysteine

  • MDL number MFCD00077103
  • Cas Number : 23032-53-3
  • Molecular Weight : 211.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209 - 211 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form Crystalline
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-benzyl-d-cysteine/2028?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-benzyl-d-cysteine/2028?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart