CH6371377957 Chem Impex International, Inc.

S-4-Methoxybenzyl-L-cysteine

  • MDL number MFCD00037207
  • Cas Number : 2544-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-217
Synonym L-Cys(pMeOBzl)-OH
Purity 99
Storage temp. Store at RT
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-4-methoxybenzyl-l-cysteine/2032?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-4-methoxybenzyl-l-cysteine/2032?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart