CH6371377959 Chem Impex International, Inc.

L-Glutamic acid a-benzyl ester

  • MDL number MFCD00037312
  • Cas Number : 13030-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Glu-OBzl
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-glutamic-acid-a-benzyl-ester/2034?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-glutamic-acid-a-benzyl-ester/2034?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart