CH6371377977 Chem Impex International, Inc.

Tosyl glycine N-hydroxysuccinimide ester

  • MDL number MFCD00153439
  • Cas Number : 68385-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Tos-Gly-OSu
Purity 99
Storage temp. Store at = -4
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/tosyl-glycine-i-n-i-hydroxysuccinimide-ester/2054?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/tosyl-glycine-i-n-i-hydroxysuccinimide-ester/2054?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart