CH6371377980 Chem Impex International, Inc.

Nalpha-Benzoyl-L-histidine methyl ester

  • MDL number MFCD00153442
  • Cas Number : 3005-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-162 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzoyl-l-histidine-methyl-ester/2058?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzoyl-l-histidine-methyl-ester/2058?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart