CH6371377986 Chem Impex International, Inc.

D-Leucine amide hydrochloride

  • MDL number MFCD00083640
  • Cas Number : 80970-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-220 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-leucine-amide-hydrochloride/2064?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-leucine-amide-hydrochloride/2064?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart