CH6371377994 Chem Impex International, Inc.

Ne-Boc-D-lysine

  • MDL number MFCD00076958
  • Cas Number : 31202-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240-246 �C
Synonym D-Lys(Boc)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-boc-d-lysine/2072?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-boc-d-lysine/2072?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart