CH6371377995 Chem Impex International, Inc.

Ne-Boc-L-lysine tert-butyl ester hydrochloride

  • MDL number MFCD00038899
  • Cas Number : 13288-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Lys(Boc)-OtBu HCl
Purity 0.98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-boc-l-lysine-i-tert-i-butyl-ester-hydrochloride/2073?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-boc-l-lysine-i-tert-i-butyl-ester-hydrochloride/2073?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart