CH6371377998 Chem Impex International, Inc.

Nalpha-Benzoyl-L-lysine

  • MDL number MFCD00038205
  • Cas Number : 366-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzoyl-l-lysine/2076?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzoyl-l-lysine/2076?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart