CH6371377999 Chem Impex International, Inc.

Na-Benzoyl-L-lysine methyl ester hydrochloride

  • MDL number MFCD00235873
  • Cas Number : 14280-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-83
Synonym Bz-L-Lys-OMe HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzoyl-l-lysine-methyl-ester-hydrochloride/2077?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzoyl-l-lysine-methyl-ester-hydrochloride/2077?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart