CH6371378001 Chem Impex International, Inc.

D-Methionine ethyl ester hydrochloride

  • MDL number MFCD00153451
  • Cas Number : 7512-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-94
Synonym D-Met-OEt HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-methionine-ethyl-ester-hydrochloride/2079?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-methionine-ethyl-ester-hydrochloride/2079?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart