CH6371378004 Chem Impex International, Inc.

Benzoyl-L-methionine

  • MDL number MFCD00066057
  • Cas Number : 10290-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-104 �C
Synonym Bz-L-Met-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzoyl-l-methionine/2082?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzoyl-l-methionine/2082?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart