CH6371378006 Chem Impex International, Inc.

Na-Benzoyl-L-ornithine

  • MDL number MFCD00153452
  • Cas Number : 17966-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-210
Synonym Bz-L-Orn-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-benzoyl-l-ornithine/2085?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-benzoyl-l-ornithine/2085?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart