CH6371378010 Chem Impex International, Inc.

D-Phenylalanine amide hydrochloride

  • MDL number MFCD00153454
  • Cas Number : 71666-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232-236 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-phenylalanine-amide-hydrochloride/2089?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-phenylalanine-amide-hydrochloride/2089?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart