CH6371378021 Chem Impex International, Inc.

Nalpha -Benzoyl-L-phenylalanine amide

  • MDL number MFCD00080883
  • Cas Number : 72150-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-206 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/n-sup-a-sup-benzoyl-l-phenylalanine-amide/2100?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-sup-a-sup-benzoyl-l-phenylalanine-amide/2100?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart