CH6371378043 Chem Impex International, Inc.

Nalpha-Benzoyl-L-tryptophan

  • MDL number MFCD00037946
  • Cas Number : 4302-66-3
  • Molecular Weight : 308.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88 - 93 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-benzoyl-l-tryptophan/2123?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-benzoyl-l-tryptophan/2123?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart