CH6371378054 Chem Impex International, Inc.

O-Benzyl-L-tyrosine benzyl ester 4-toluenesulfonate salt

  • MDL number MFCD00077105
  • Cas Number : 66009-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Tyr(Bzl)-OBzl TosOH
Storage temp. Store at 0-8
Physical Form Crystalline
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-benzyl-l-tyrosine-benzyl-ester-4-toluenesulfonate-salt/2135?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-benzyl-l-tyrosine-benzyl-ester-4-toluenesulfonate-salt/2135?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart