CH6371378064 Chem Impex International, Inc.

Fmoc-L-alaninol

  • MDL number MFCD00235913
  • Cas Number : 161529-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-157
Synonym Fmoc-L-Ala-ol
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-alaninol/2145?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-alaninol/2145?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart