CH6371378069 Chem Impex International, Inc.

Fmoc-L-asparaginol

  • MDL number MFCD00235916
  • Cas Number : 260450-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-181° C
Purity 97
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-asparaginol/2150?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-asparaginol/2150?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart