CH6371378071 Chem Impex International, Inc.

S-4-methylbenzyl-L-cysteinol

  • MDL number MFCD00671686
  • Cas Number : 438237-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51 - 56 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-4-methylbenzyl-l-cysteinol/2152?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-4-methylbenzyl-l-cysteinol/2152?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart