CH6371378073 Chem Impex International, Inc.

Boc-S-4-methylbenzyl-D-cysteinol

  • MDL number MFCD00235920

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-70° C
Purity 99
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-d-cysteinol/2154?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-d-cysteinol/2154?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart