CH6371378074 Chem Impex International, Inc.

S-benzyl-L-cysteinol

  • MDL number MFCD00235921
  • Cas Number : 85803-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Cysteinol(Bzl)
Purity 99.5
Storage temp. Store at 0-8
Physical Form White to tan powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-benzyl-l-cysteinol/2155?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-benzyl-l-cysteinol/2155?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart