CH6371378075 Chem Impex International, Inc.

Fmoc-S-acetamidomethyl-L-cysteinol

  • MDL number MFCD02094116
  • Cas Number : 198543-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Cysteinol(Acm)
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-acetamidomethyl-l-cysteinol/2156?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-acetamidomethyl-l-cysteinol/2156?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart