CH6371378076 Chem Impex International, Inc.

Boc-S-benzyl-L-cysteinol

  • MDL number MFCD00235923
  • Cas Number : 139428-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66 - 70 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-benzyl-l-cysteinol/2157?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-benzyl-l-cysteinol/2157?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart