CH6371378077 Chem Impex International, Inc.

Boc-S-benzyl-D-cysteinol

  • MDL number MFCD00235924
  • Cas Number : 198470-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-70°C
Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-benzyl-d-cysteinol/2158?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-benzyl-d-cysteinol/2158?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart