CH6371378078 Chem Impex International, Inc.

Fmoc-L-glutaminol

  • MDL number MFCD00235925
  • Cas Number : 260450-77-9
  • Molecular Weight : 354.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-197° C
Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-glutaminol/2159?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-glutaminol/2159?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart