CH6371378083 Chem Impex International, Inc.

L-Isoleucinol hydrochloride

  • MDL number MFCD00038956
  • Cas Number : 133736-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-136 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-isoleucinol-hydrochloride/2164?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-isoleucinol-hydrochloride/2164?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart