CH6371378090 Chem Impex International, Inc.

Fmoc-L-leucinol

  • MDL number MFCD00235932
  • Cas Number : 139551-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-134
Synonym Fmoc-Leu-ol
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-leucinol/2171?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-leucinol/2171?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart