CH6371378091 Chem Impex International, Inc.

Nepsilon-Z-L-lysinol

  • MDL number MFCD00671688
  • Cas Number : 101250-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-z-l-lysinol/2172?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-z-l-lysinol/2172?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart