CH6371378092 Chem Impex International, Inc.

Na-Boc-Ne-Z-L-lysinol

  • MDL number MFCD00235934
  • Cas Number : 82689-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Lysinol(Z)
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-z-l-lysinol/2173?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-z-l-lysinol/2173?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart