CH6371378093 Chem Impex International, Inc.

Na-Boc-Ne-Z-D-lysinol

  • MDL number MFCD00235935
  • Cas Number : 252940-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-68� C
Synonym Boc-D-Lysinol(Z)
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-z-d-lysinol/2174?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-z-d-lysinol/2174?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart