CH6371378094 Chem Impex International, Inc.

Nalpha-Boc-Nepsilon-2-chloro-Z-L-lysinol

  • MDL number MFCD00235936
  • Cas Number : 198476-84-5
  • Molecular Weight : 400.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71 - 72 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-2-chloro-z-l-lysinol/2175?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-2-chloro-z-l-lysinol/2175?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart