CH6371378095 Chem Impex International, Inc.

Na-Fmoc-Ne-Boc-L-lysinol

  • MDL number MFCD00235937
  • Cas Number : 198561-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-142� C (Dec)
Synonym Fmoc-L-Lysinol(Boc)
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-e-sup-boc-l-lysinol/2176?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-e-sup-boc-l-lysinol/2176?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart