CH6371378098 Chem Impex International, Inc.

Boc-L-methioninol

  • MDL number MFCD00235939
  • Cas Number : 51372-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-Met-ol
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-methioninol/2179?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-methioninol/2179?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart