CH6371378099 Chem Impex International, Inc.

Boc-D-Methioninol

  • MDL number MFCD00235940
  • Cas Number : 91177-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-53
Synonym Boc-D-Met-ol
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-methioninol/2180?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-methioninol/2180?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart