CH6371378117 Chem Impex International, Inc.

Boc-O-benzyl-D-threoninol

  • MDL number MFCD00235949
  • Cas Number : 168034-31-9
  • Molecular Weight : 295.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form Oil
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-threoninol/2198?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-threoninol/2198?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart