CH6371378118 Chem Impex International, Inc.

Fmoc-O-tert-butyl-L-threoninol

  • MDL number MFCD00235950
  • Cas Number : 189337-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Threoninol(tBu)
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-i-tert-i-butyl-l-threoninol/2199?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-i-tert-i-butyl-l-threoninol/2199?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart