CH6371378123 Chem Impex International, Inc.

Tyrosinol(benzyl)

  • MDL number MFCD00235955
  • Cas Number : 85803-44-7
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/tyrosinol-benzyl/2204?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/tyrosinol-benzyl/2204?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart