CH6371378126 Chem Impex International, Inc.

Boc-O-2,6-dichlorobenzyl-L-tyrosinol

  • MDL number MFCD00235958
  • Cas Number : 169393-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Tyrosinol(2,6-di-Cl-Bzl)
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-2-6-dichlorobenzyl-l-tyrosinol/2207?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-2-6-dichlorobenzyl-l-tyrosinol/2207?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart