CH6371378127 Chem Impex International, Inc.

Fmoc-O-tert-butyl-L-tyrosinol

  • MDL number MFCD00235959
  • Cas Number : 187526-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-115 �C
Synonym Fmoc-L-Tyrosinol(tBu)
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-i-tert-i-butyl-l-tyrosinol/2208?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-i-tert-i-butyl-l-tyrosinol/2208?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart