CH6371378136 Chem Impex International, Inc.

Ac-Phe-3,5-diI-Tyr-OH

  • MDL number MFCD00002180
  • Cas Number : 3786-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ac-phe-3-5-dii-tyr-oh/2223?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ac-phe-3-5-dii-tyr-oh/2223?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart