CH6371378137 Chem Impex International, Inc.

Acetyl-L-threonine

  • MDL number MFCD00037810
  • Cas Number : 17093-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-L-Thr-OH
Purity 97
Storage temp. Store at 0-8
Physical Form Stickey solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-threonine/2224?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-threonine/2224?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart